Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-1H-pyrrolo[3,2-b]pyridine 97%

Supplier: AMBEED, INC
Warning

A596961-100MG A596961-1G A596961-250MG
77197-954EA 17 USD InStock
77197-954 77197-956 77197-958
1-Methyl-1H-pyrrolo[3,2-b]pyridine 97%
1-Methyl-1H-pyrrolo[3,2-b]pyridine
Precaución: for Research Use Only


Formula: C₈H₈N₂
MDL Number: MFCD12923567
CAS Number: 153374-33-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty