Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,4-Phenylene diacrylate 97%

Supplier: AMBEED, INC
Warning

A598197-100MG A598197-1G A598197-250MG
77197-972EA 20 USD InStock
77197-972 77197-974 77197-976
1,4-Phenylene diacrylate 97%
1,4-Phenylene diacrylate
Precaución: for Research Use Only


Formula: C₁₂H₁₀O₄
MW: 218.20 g/mol
MDL Number: MFCD00080618
CAS Number: 6729-79-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty