Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Chlorophenyl)-4,4,4-trifluoro-1,3-butanedione 98%

Supplier: AMBEED, INC
Warning

A601531-1G A601531-25G A601531-5G A601531-10G
77198-026EA 11 USD InStock
77198-026 77198-028 77198-030 77747-244
1-(4-Chlorophenyl)-4,4,4-trifluoro-1,3-butanedione 98%
1-(4-Chlorophenyl)-4,4,4-trifluoro-1,3-butanedione
Precaución: for Research Use Only


Formula: C₁₀H₆ClF₃O₂
MDL Number: MFCD00511276
CAS Number: 18931-60-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty