Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

P529 98+%

Supplier: AMBEED, INC
Warning

A602029-10MG A602029-1MG A602029-50MG A602029-5MG
77170-118EA 43 USD InStock
77170-118 77170-120 77170-122 77170-124
P529 98+%
P529
Precaución: for Research Use Only


Formula: C₂₄H₂₂O₆
MW: 406.43 g/mol
MDL Number: MFCD18633224
CAS Number: 914913-88-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty