Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-(Benzyloxy)-[1,1'-biphenyl]-4-carboxylic acid 98%

Supplier: AMBEED, INC
Warning

A609626-100MG A609626-1G A609626-250MG
77198-100EA 11 USD InStock
77198-100 77198-102 77198-104
4'-(Benzyloxy)-[1,1'-biphenyl]-4-carboxylic acid 98%
4'-(Benzyloxy)-[1,1'-biphenyl]-4-carboxylic acid
Precaución: for Research Use Only


Formula: C₂₀H₁₆O₃
MW: 304.33 g/mol
MDL Number: MFCD06802253
CAS Number: 111153-16-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty