Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(2-Methoxyethoxy)propanoic acid 95%, Ambeed.Inc

Supplier: AMBEED, INC
Danger

A609900-250MG A609900-100MG
77170-194EA 47 USD InStock
77170-194 77684-484
3-(2-Methoxyethoxy)propanoic acid 95%, Ambeed.Inc
3-(2-Methoxyethoxy)propanoic acid
Precaución: For research use only.


Formula: C₆H₁₂O₄
MW: 148.157 g/mol
MDL Number: MFCD09939037
CAS Number: 149577-05-9
UN: 3265
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty