Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-1-(methylsulfonyl)indoline 98%

Supplier: AMBEED, INC
Warning

A610762-100MG A610762-1G A610762-250MG
77198-120EA 5 USD InStock
77198-120 77198-122 77198-124
5-Bromo-1-(methylsulfonyl)indoline 98%
5-Bromo-1-(methylsulfonyl)indoline
Precaución: for Research Use Only


Formula: C₉H₁₀BrNO₂S
MDL Number: MFCD02853099
CAS Number: 446054-18-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty