Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-Methyl 2,3-diaminopropanoate dihydrochloride 97%

Supplier: AMBEED, INC
Warning

A613913-250MG A613913-5G
77198-140EA 39 USD InStock
77198-140 77198-142
(S)-Methyl 2,3-diaminopropanoate dihydrochloride 97%
(S)-Methyl 2,3-diaminopropanoate dihydrochloride
Precaución: for Research Use Only


Formula: C₄H₁₀N₂O₂·2ClH
MDL Number: MFCD09998031
CAS Number: 147857-43-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty