Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Morpholino-3-(morpholinosulfonyl)aniline 97%

Supplier: AMBEED, INC
Warning

A617837-100MG A617837-1G A617837-250MG
77198-180EA 22 USD InStock
77198-180 77198-182 77198-184
4-Morpholino-3-(morpholinosulfonyl)aniline 97%
4-Morpholino-3-(morpholinosulfonyl)aniline
Precaución: for Research Use Only


Formula: C₁₄H₂₁N₃O₄S
MDL Number: MFCD00087613
CAS Number: 200711-95-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty