Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-[2-(2-Amino-4-chlorophenoxy)ethyl]-N,N-dimethylamine 97%

Supplier: AMBEED, INC
Warning

A618083-1G A618083-250MG A618083-5G
77198-186EA 105 USD InStock
77198-186 77198-188 77198-190
N-[2-(2-Amino-4-chlorophenoxy)ethyl]-N,N-dimethylamine 97%
N-[2-(2-Amino-4-chlorophenoxy)ethyl]-N,N-dimethylamine
Precaución: for Research Use Only


Formula: C₁₀H₁₅ClN₂O
MDL Number: MFCD08686778
CAS Number: 631862-75-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty