Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dichloro-2-methylpyrazine 98%

Supplier: AMBEED, INC
Warning

A619342-100MG A619342-1G A619342-250MG
77170-206EA 35 USD InStock
77170-206 77170-208 77170-210
3,5-Dichloro-2-methylpyrazine 98%
3,5-Dichloro-2-methylpyrazine
Precaución: for Research Use Only


Formula: C₅H₄Cl₂N₂
MDL Number: MFCD12024888
CAS Number: 89284-38-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty