Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,14-Dibromotetradecane 98%

Supplier: AMBEED, INC
Warning

A634332-100MG A634332-1G A634332-250MG A634332-5G A634332-25G A634332-100G
77198-302EA 22 USD InStock
77198-302 77198-304 77198-306 77198-308 77747-308 77747-307
1,14-Dibromotetradecane 98%
1,14-Dibromotetradecane
Precaución: for Research Use Only


Formula: BrH₂C(CH₂)₁₂CH₂Br
MDL Number: MFCD00045049
CAS Number: 37688-96-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty