Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-6-(trifluoromethyl)benzoyl chloride 98%

Supplier: AMBEED, INC
Danger

A637629-25G A637629-5G
77198-322EA 178 USD InStock
77198-322 77198-324
2-Fluoro-6-(trifluoromethyl)benzoyl chloride 98%
2-Fluoro-6-(trifluoromethyl)benzoyl chloride
Precaución: for Research Use Only


Formula: C₈H₃ClF₄O
MW: 226.56
Density: 1.465
MDL Number: MFCD00061160
CAS Number: 109227-12-5
UN: 3265
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty