Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-bromo-3-fluoro-2-methylbenzoate 97%

Supplier: AMBEED, INC
Warning

A639606-100MG A639606-1G A639606-250MG
77198-348EA 29 USD InStock
77198-348 77198-350 77198-352
Methyl 4-bromo-3-fluoro-2-methylbenzoate 97%
Methyl 4-bromo-3-fluoro-2-methylbenzoate
Precaución: for Research Use Only


Formula: C₉H₈BrFO₂
MW: 247.06 g/mol
MDL Number: MFCD21602051
CAS Number: 1365969-22-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty