Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1-Aminocyclobutyl)methanol hydrochloride 97%

Supplier: AMBEED, INC
Warning

A654374-100MG A654374-1G A654374-250MG A654374-5G
77170-428EA 10 USD InStock
77170-428 77170-430 77170-432 77170-434
(1-Aminocyclobutyl)methanol hydrochloride 97%
(1-Aminocyclobutyl)methanol hydrochloride
Precaución: for Research Use Only


Formula: C₅H₁₁NO·ClH
MDL Number: MFCD17015024
CAS Number: 1392213-15-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty