Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Dimethyl 4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phthalate 97%

Supplier: AMBEED, INC
Warning

A658384-100MG A658384-1G A658384-250MG
77198-480EA 39 USD InStock
77198-480 77198-482 77198-484
1,2-Dimethyl 4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phthalate 97%
1,2-Dimethyl 4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phthalate


Formula: C₁₆H₂₁BO₆
MDL Number: MFCD28397295
CAS Number: 2157414-14-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty