Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromophthalic acid 98%

Supplier: AMBEED, INC
Warning

A659183-100G A659183-1G A659183-25G A659183-5G
77198-504EA 180 USD InStock
77198-504 77198-506 77198-508 77198-510
4-Bromophthalic acid 98%
4-Bromophthalic acid
Precaución: for Research Use Only


Formula: C₈H₅BrO₄
MW: 245.03 g/mol
MDL Number: MFCD00043538
CAS Number: 6968-28-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty