Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-3,3'-Bis(3,5-dichlorophenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate 98%

Supplier: AMBEED, INC
Warning

A662094-100MG A662094-250MG
77198-528EA 144 USD InStock
77198-528 77198-530
(R)-3,3'-Bis(3,5-dichlorophenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate 98%
(R)-3,3'-Bis(3,5-dichlorophenyl)-1,1'-binapthyl-2,2'-diyl hydrogenphosphate
Precaución: for Research Use Only


Formula: C₃₂H₁₇Cl₄O₄P
MDL Number: MFCD23698720
CAS Number: 1191451-24-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty