Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-7-fluoropyrrolo[1,2-a]quinoxaline 98%

Supplier: AMBEED, INC
Warning

A665802-100MG A665802-1G A665802-250MG
77170-504EA 39 USD InStock
77170-504 77170-506 77170-508
4-Chloro-7-fluoropyrrolo[1,2-a]quinoxaline 98%
4-Chloro-7-fluoropyrrolo[1,2-a]quinoxaline
Precaución: for Research Use Only


Formula: C₁₁H₆ClFN₂
MDL Number: MFCD09835554
CAS Number: 136773-69-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty