Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

RO 48-8071 98%

Supplier: AMBEED, INC
Warning

A674342-10MG A674342-1MG A674342-25MG A674342-50MG A674342-5MG
77170-552EA 57 USD InStock
77170-552 77170-554 77170-556 77170-558 77170-560
RO 48-8071 98%
RO 48-8071
Precaución: for Research Use Only


Formula: C₂₃H₂₇BrFNO₂.C₄H₄O₄
MW: 564.44 g/mol
MDL Number: MFCD05865242
CAS Number: 189197-69-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty