Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2-Dimethyl-1,3-dioxacyclohept-5-ene 98% (by GC)

Supplier: AMBEED, INC
Warning

A688510-100G A688510-1G A688510-25G A688510-5G
77170-626EA 304 USD InStock
77170-626 77170-628 77170-630 77170-632
2,2-Dimethyl-1,3-dioxacyclohept-5-ene 98% (by GC)
2,2-Dimethyl-1,3-dioxacyclohept-5-ene
Precaución: for Research Use Only


Formula: C₇H₁₂O₂
MDL Number: MFCD22054122
CAS Number: 1003-83-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty