Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-7-methoxy-1-methyl-1H-benzo[d][1,2,3]triazole 97%

Supplier: AMBEED, INC
Warning

A692371-100MG A692371-1G A692371-250MG
77198-738EA 50 USD InStock
77198-738 77198-740 77198-742
5-Bromo-7-methoxy-1-methyl-1H-benzo[d][1,2,3]triazole 97%
5-Bromo-7-methoxy-1-methyl-1H-benzo[d][1,2,3]triazole
Precaución: for Research Use Only


Formula: C₈H₈BrN₃O
MDL Number: MFCD28969711
CAS Number: 1799973-82-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty