Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4''-Ethyl-2'-fluoro-4-propyl-1,1':4',1''-terphenyl 97%

Supplier: AMBEED, INC
Warning

A692579-25G A692579-5G
77198-744EA 66 USD InStock
77198-744 77198-746
4''-Ethyl-2'-fluoro-4-propyl-1,1':4',1''-terphenyl 97%
4''-Ethyl-2'-fluoro-4-propyl-1,1':4',1''-terphenyl
Precaución: for Research Use Only


Formula: C₂₃H₂₃F
MDL Number: MFCD01830216
CAS Number: 95759-44-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty