Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyridine-2-carboxaldoxime 98%

Supplier: AMBEED, INC
Danger

A702070-100G A702070-25G A702070-5G
77170-714EA 209 USD InStock
77170-714 77170-716 77170-718
Pyridine-2-carboxaldoxime 98%
Pyridine-2-carboxaldoxime
Precaución: for Research Use Only


Formula: C₆H₆N₂O
MDL Number: MFCD00006354
CAS Number: 873-69-8
UN: 1325
ADR: 4.1,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty