Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(-)-Tetrahydro-2-furoic acid 97%

Supplier: AMBEED, INC
Danger

A711273-10G A711273-25G A711273-5G
77170-798EA 14 USD InStock
77170-798 77170-800 77170-802
(S)-(-)-Tetrahydro-2-furoic acid 97%
(S)-(-)-Tetrahydro-2-furoic acid
Precaución: for Research Use Only


Formula: C₅H₈O₃
MDL Number: MFCD00272639
CAS Number: 87392-07-2
UN: 3265
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty