Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 6-(trifluoromethyl)-1H-indazole-3-carboxylate 98%

Supplier: AMBEED, INC
Warning

A716514-100MG A716514-1G A716514-250MG
77170-820EA 17 USD InStock
77170-820 77170-822 77170-824
Methyl 6-(trifluoromethyl)-1H-indazole-3-carboxylate 98%
Methyl 6-(trifluoromethyl)-1H-indazole-3-carboxylate
Precaución: for Research Use Only


Formula: C₁₀H₇F₃N₂O₂
MDL Number: MFCD09878903
CAS Number: 877773-17-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty