Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Dihydro-4-oxo-1,2,3-benzotriazine 98%

Supplier: AMBEED, INC
Warning

A716518-100G A716518-1G A716518-25G A716518-5G
77198-954EA 194 USD InStock
77198-954 77198-956 77198-958 77198-960
3,4-Dihydro-4-oxo-1,2,3-benzotriazine 98%
3,4-Dihydro-4-oxo-1,2,3-benzotriazine
Precaución: for Research Use Only


Formula: C₇H₅N₃O
MDL Number: MFCD00052387
CAS Number: 90-16-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty