Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethylenedithiobis(trimethylsilane) 97%

Supplier: AMBEED, INC
Warning

A724321-1G A724321-250MG A724321-25G A724321-5G
77199-026EA 21 USD InStock
77199-026 77199-028 77199-030 77199-032
Ethylenedithiobis(trimethylsilane) 97%
Ethylenedithiobis(trimethylsilane)
Precaución: for Research Use Only


Formula: C₈H₂₂S₂Si₂
MDL Number: MFCD00042832
CAS Number: 51048-29-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty