Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Piperidin-4-ylidene)acetic acid hydrochloride 97%

Supplier: AMBEED, INC
Warning

A734237-100MG A734237-1G A734237-250MG
77170-946EA 8 USD InStock
77170-946 77170-948 77170-950
2-(Piperidin-4-ylidene)acetic acid hydrochloride 97%
2-(Piperidin-4-ylidene)acetic acid hydrochloride
Precaución: for Research Use Only


Formula: C₇H₁₂ClNO₂
MDL Number: MFCD11506216
CAS Number: 84839-57-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty