Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(6-Methylpyridin-2-yl)morpholine 95%

Supplier: AMBEED, INC
Warning

A738224-1G A738224-250MG A738224-5G
77199-128EA 40 USD InStock
77199-128 77199-130 77199-132
4-(6-Methylpyridin-2-yl)morpholine 95%
4-(6-Methylpyridin-2-yl)morpholine
Precaución: for Research Use Only


Formula: C₁₀H₁₄N₂O
MDL Number: MFCD28369599
CAS Number: 1321517-90-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty