Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-3-methyl-5-nitroaniline 95%

Supplier: AMBEED, INC
Warning

A744138-100MG A744138-250MG
77170-996EA 32 USD InStock
77170-996 77170-998
2-Bromo-3-methyl-5-nitroaniline 95%
2-Bromo-3-methyl-5-nitroaniline
Precaución: for Research Use Only


Formula: C₇H₇BrN₂O₂
MDL Number: MFCD28099606
CAS Number: 631911-97-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty