Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyldimethyl(2-propynyloxy)silane 98%

Supplier: AMBEED, INC
Danger

A745693-25G A745693-5G
77171-000EA 16 USD InStock
77171-000 77171-002
tert-Butyldimethyl(2-propynyloxy)silane 98%
tert-Butyldimethyl(2-propynyloxy)silane
Precaución: for Research Use Only


Formula: C₉H₁₈OSi
MDL Number: MFCD28156589
CAS Number: 76782-82-6
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty