Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Piperidin-1-yl)acetic acid hydrate 97%

Supplier: AMBEED, INC
Warning

A748771-1G A748771-250MG
77199-204EA 11 USD InStock
77199-204 77199-206
2-(Piperidin-1-yl)acetic acid hydrate 97%
2-(Piperidin-1-yl)acetic acid hydrate
Precaución: for Research Use Only


Formula: C₇H₁₅NO₃
MW: 161.2 g/mol
MDL Number: MFCD07781088
CAS Number: 478920-83-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty