Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N5-(6-Aminohexyl)-N7-benzyl-3-isopropylpyrazolo[1,5-a]pyrimidine-5,7-diamine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A750865-10MG A750865-1MG A750865-2MG A750865-50MG A750865-5MG
77199-216EA 70 USD InStock
77199-216 77199-218 77199-220 77199-222 77199-224
N5-(6-Aminohexyl)-N7-benzyl-3-isopropylpyrazolo[1,5-a]pyrimidine-5,7-diamine hydrochloride 98%
N5-(6-Aminohexyl)-N7-benzyl-3-isopropylpyrazolo[1,5-a]pyrimidine-5,7-diamine hydrochloride
Precaución: for Research Use Only


Formula: C₂₂H₃₃ClN₆
MDL Number: MFCD18384975
CAS Number: 1397219-81-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty