Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Pyrrolo[3,2-c]pyridine-2-carbaldehyde 98%

Supplier: AMBEED, INC
Warning

A751629-100MG A751629-250MG A751629-5G
77171-064EA 23 USD InStock
77171-064 77171-066 77171-068
1H-Pyrrolo[3,2-c]pyridine-2-carbaldehyde 98%
1H-Pyrrolo[3,2-c]pyridine-2-carbaldehyde
Precaución: for Research Use Only


Formula: C₈H₆N₂O
MW: 146.15 g/mol
MDL Number: MFCD06738310
CAS Number: 630395-95-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty