Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Z)-N'-hydroxy-4-methylbenzimidamide 98%

Supplier: AMBEED, INC
Danger

A757732-1G A757732-5G
77199-258EA 13 USD InStock
77199-258 77199-260
(Z)-N'-hydroxy-4-methylbenzimidamide 98%
(Z)-N'-hydroxy-4-methylbenzimidamide
Precaución: for Research Use Only


Formula: C₈H₁₀N₂O
MW: 150.17 g/mol
MDL Number: MFCD00063016
CAS Number: 923023-83-0
UN: 1325
ADR: 4.1 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty