Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-Methyl-1,3-thiazol-4-yl)ethanone 96%

Supplier: AMBEED, INC
Warning

A758248-1G A758248-250MG
77199-264EA 39 USD InStock
77199-264 77199-266
1-(2-Methyl-1,3-thiazol-4-yl)ethanone 96%
1-(2-Methyl-1,3-thiazol-4-yl)ethanone
Precaución: for Research Use Only


Formula: C₆H₇NOS
MDL Number: MFCD00278580
CAS Number: 23002-78-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty