Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Iodo-o-toluidine 98%

Supplier: AMBEED, INC
Warning

A758914-100G A758914-10G A758914-1G A758914-250MG A758914-25G A758914-5G
77199-278EA 95 USD InStock
77199-278 77199-280 77199-282 77199-284 77199-286 77199-288
5-Iodo-o-toluidine 98%
5-Iodo-o-toluidine
Precaución: for Research Use Only


Formula: C₇H₈IN
MDL Number: MFCD00051531
CAS Number: 83863-33-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty