Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Indole-2-sulfonamide 97%

Supplier: AMBEED, INC
Warning

A764158-250MG A764158-50MG
77199-316EA 641 USD InStock
77199-316 77199-318
1H-Indole-2-sulfonamide 97%
1H-Indole-2-sulfonamide
Precaución: for Research Use Only


Formula: C₈H₈N₂O₂S
MW: 196.22 g/mol
MDL Number: MFCD18809113
CAS Number: 85953-41-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty