Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

PIK-93 ≥98%

Supplier: AMBEED, INC
Warning

A764324-10MG A764324-1MG A764324-50MG A764324-5MG A764324-100MG A764324-250MG
77199-328EA 66 USD InStock
77199-328 77199-330 77199-332 77199-334 77747-509 77747-510
PIK-93 ≥98%
PIK-93
Precaución: for Research Use Only


Formula: C₁₄H₁₆ClN₃O₄S₂
MDL Number: MFCD12922510
CAS Number: 593960-11-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty