Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-α-methylbenzyl alcohol 98%

Supplier: AMBEED, INC
Warning

A765144-1G A765144-25G A765144-5G
77199-346EA 11 USD InStock
77199-346 77199-348 77199-350
4-Amino-α-methylbenzyl alcohol 98%
4-Amino-α-methylbenzyl alcohol
Precaución: for Research Use Only


Formula: C₈H₁₁NO
MW: 137.18 g/mol
MDL Number: MFCD00014781
CAS Number: 14572-89-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty