Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Chloro-1-iodo-3-isopropylimidazo[1,5-a]pyrazine 95%

Supplier: AMBEED, INC
Warning

A774224-100MG A774224-1G A774224-250MG
77199-416EA 96 USD InStock
77199-416 77199-418 77199-420
8-Chloro-1-iodo-3-isopropylimidazo[1,5-a]pyrazine 95%
8-Chloro-1-iodo-3-isopropylimidazo[1,5-a]pyrazine
Precaución: for Research Use Only


Formula: C₉H₉ClIN₃
MW: 321.55 g/mol
MDL Number: MFCD20489126
CAS Number: 1320266-92-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty