Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(3-Chlorophenyl)-1H-imidazole-5-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A776469-100MG A776469-250MG
77199-430EA 88 USD InStock
77199-430 77199-432
1-(3-Chlorophenyl)-1H-imidazole-5-carboxylic acid 97%
1-(3-Chlorophenyl)-1H-imidazole-5-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₀H₇ClN₂O₂
MDL Number: MFCD25368754
CAS Number: 1197398-96-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty