Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4-(Cyclohexane-1,1-diyl)bis(N,N-di-p-tolylaniline) 98%

Supplier: AMBEED, INC
Warning

A781065-100MG A781065-1G A781065-250MG A781065-5G
77199-444EA 10 USD InStock
77199-444 77199-446 77199-448 77199-450
4,4-(Cyclohexane-1,1-diyl)bis(N,N-di-p-tolylaniline) 98%
4,4-(Cyclohexane-1,1-diyl)bis(N,N-di-p-tolylaniline)
Precaución: for Research Use Only


Formula: C₄₆H₄₆N₂
MDL Number: MFCD01310666
CAS Number: 58473-78-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty