Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(3-Bromobenzyl)pyrrolidine 95%

Supplier: AMBEED, INC
Warning

A781704-10G A781704-1G A781704-250MG A781704-25G
77171-234EA 45 USD InStock
77171-234 77171-236 77171-238 77171-240
1-(3-Bromobenzyl)pyrrolidine 95%
1-(3-Bromobenzyl)pyrrolidine
Precaución: for Research Use Only


Formula: C₁₁H₁₄BrN
MW: 240.15 g/mol
MDL Number: MFCD06080721
CAS Number: 168820-15-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty