Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Difluoro-3-(trifluoromethyl)benzoic acid 98%

Supplier: AMBEED, INC
Warning

A784514-100MG A784514-1G A784514-250MG
77171-274EA 18 USD InStock
77171-274 77171-276 77171-278
2,6-Difluoro-3-(trifluoromethyl)benzoic acid 98%
2,6-Difluoro-3-(trifluoromethyl)benzoic acid
Precaución: for Research Use Only


Formula: C₈H₃F₅O₂
MW: 226.1 g/mol
MDL Number: MFCD03412249
CAS Number: 1048921-49-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty