Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Chlorophenyl)-3-methyl-N-(thiazol-2-yl)butanamide 97%

Supplier: AMBEED, INC
Warning

A789179-50MG A789179-250MG A789179-100MG
77199-520EA 58 USD InStock
77199-520 77747-537 77747-536
2-(4-Chlorophenyl)-3-methyl-N-(thiazol-2-yl)butanamide 97%
2-(4-Chlorophenyl)-3-methyl-N-(thiazol-2-yl)butanamide
Precaución: for Research Use Only


Formula: C₁₄H₁₅ClN₂OS
MDL Number: MFCD02666163
CAS Number: 300851-67-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty