Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Diphenyl-1H-pyrazol-5-amine 95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A801112-1G A801112-250MG A801112-5G A801112-100MG
77171-372EA 27 USD InStock
77171-372 77171-374 77171-376 77684-746
1,3-Diphenyl-1H-pyrazol-5-amine 95%, Ambeed.Inc
1,3-Diphenyl-1H-pyrazol-5-amine
Precaución: For research use only.


Formula: C₁₅H₁₃N₃
MW: 235.29 g/mol
MDL Number: MFCD00084878
CAS Number: 5356-71-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty