Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl (S)-2-isocyanato-3-phenylpropionate 97%

Supplier: AMBEED, INC
Danger

A807096-100MG A807096-1G A807096-5G
77199-652EA 17 USD InStock
77199-652 77199-654 77199-656
Methyl (S)-2-isocyanato-3-phenylpropionate 97%
Methyl (S)-2-isocyanato-3-phenylpropionate
Precaución: for Research Use Only


Formula: C₁₁H₁₁NO₃
MDL Number: MFCD00191535
CAS Number: 40203-94-9
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty