Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Aminosalicylic acid 95%

Supplier: AMBEED, INC
Warning

A810773-1G A810773-5G A810773-10G
77199-716EA 55 USD InStock
77199-716 77199-718 77747-561
3-Aminosalicylic acid 95%
3-Aminosalicylic acid
Precaución: for Research Use Only


Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 347
Melting Pt: 290
MDL Number: MFCD00010299
CAS Number: 570-23-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty